Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201365.1 | NP_001188294.1 | 5 | 777 | ||
NM_004290.4 | NP_004281.1 | 5 | 991 | |||
NM_183398.2 | NP_899645.1 | 4 | 839 | |||
NM_183399.2 | NP_899646.1 | 4 | 603 | |||
NM_183400.2 | NP_899647.1 | 4 | 817 | |||
NM_183401.2 | NP_899648.1 | 5 | 797 | |||
XM_005268536.2 | XP_005268593.1 | 5 | 1094 | |||
XM_005268537.2 | XP_005268594.1 | 5 | 1138 | |||
XM_005268539.2 | XP_005268596.1 | 4 | 607 | |||
XM_005268540.3 | XP_005268597.1 | 4 | 763 | |||
XM_005268541.3 | XP_005268598.1 | 3 | 519 | |||
XM_011537714.2 | XP_011536016.1 | 4 | 608 | |||
XM_017010070.1 | XP_016865559.1 | 5 | 742 | |||
XM_017010071.1 | XP_016865560.1 | 4 | 969 | |||
XM_017010072.1 | XP_016865561.1 | 4 | 929 | |||
GenBank mRNA | AB022663.1 | - | 5 | 765 | ||
AF060544.1 | - | 4 | 568 | |||
AK023698.1 | - | 4 | 567 | |||
AK023884.1 | - | 5 | 733 | |||
AK057868.1 | - | 4 | 615 | |||
AK129515.1 | - | 4 | 768 | |||
BC126185.1 | - | 5 | 736 | 8991901 | ||
BC144061.1 | - | 5 | 736 | 9052581 | ||
BX647955.1 | - | 5 | 764 | |||
CD106708.1 | - | 5 | 802 | |||
CR456702.1 | - | 3 | 529 | |||
DB456366.2 | - | 4 | 682 |