Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253770.2 | XP_006253832.1 | 9 | 1061 | ||
GenBank mRNA | BC158556.1 | - | 9 | 967 | 9030831 |