Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099519.1 | NP_001092989.1 | 3 | 633 |
Set Membership: |
G-protein coupled receptor |