Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001073777.4 | XP_001073777.2 | 14, 15 | 1841 | ||
XM_006244268.2 | XP_006244330.1 | 15, 16 | 1907 | |||
XM_006244269.2 | XP_006244331.1 | 15, 16 | 2136 | |||
XM_006244270.2 | XP_006244332.1 | 15, 16 | 2421 | |||
XM_006244271.2 | XP_006244333.1 | 15, 16 | 2433 | |||
XM_008766793.1 | XP_008765015.1 | 14, 15 | 2266 | |||
XM_008766794.1 | XP_008765016.1 | 11, 12 | 2525 |