Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004291.3 | NP_004282.1 | 3 | 804 | ||
GenBank mRNA | U16826.1 | - | 3 | 687 |