Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC681871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759288.1 | XP_008757510.1 | 3 | 554 |
Gene Symbol: Vom2r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099463.1 | NP_001092933.1 | 554 |
Set Membership: |
G-protein coupled receptor |