Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257418.2 | XP_006257480.1 | 2 | 3006 | ||
XM_008773478.1 | XP_008771700.1 | 5 | 2377 | |||
XM_008773479.1 | XP_008771701.1 | 3 | 3209 | |||
XM_008773480.1 | XP_008771702.1 | 2 | 707 |