Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTRF1 SIRNA Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
UniGene ID:
Hs.382176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18478
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTRF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004294.2
NP_004285.2
9
1256
XM_005266599.2
XP_005266656.1
11
1505
XM_006719897.2
XP_006719960.1
11
1613
XM_006719898.2
XP_006719961.1
11
1597
XM_006719900.2
XP_006719963.1
10
1326
XM_011535315.2
XP_011533617.1
11
1722
XM_011535317.2
XP_011533619.1
10
1605
XM_011535318.2
XP_011533620.1
11
1765
XM_011535320.2
XP_011533622.1
11
1701
XM_011535321.2
XP_011533623.1
14
1837
XM_011535323.2
XP_011533625.1
11
1567
XM_011535325.2
XP_011533627.1
10
1520
XM_017020856.1
XP_016876345.1
10
1566
XM_017020857.1
XP_016876346.1
13
1700
XM_017020858.1
XP_016876347.1
12
1695
XM_017020859.1
XP_016876348.1
10
1566
XM_017020860.1
XP_016876349.1
12
1583
XM_017020861.1
XP_016876350.1
12
1620
XM_017020862.1
XP_016876351.1
11
2778
XM_017020863.1
XP_016876352.1
11
2626
XM_017020864.1
XP_016876353.1
11
2518
XM_017020865.1
XP_016876354.1
10
2470
XM_017020866.1
XP_016876355.1
9
2335
XM_017020867.1
XP_016876356.1
11
1483
XM_017020868.1
XP_016876357.1
9
1451
XM_017020869.1
XP_016876358.1
10
1349
XM_017020870.1
XP_016876359.1
11
1613
XM_017020871.1
XP_016876360.1
10
1326
XM_017020873.1
XP_016876362.1
14
1907
XM_017020874.1
XP_016876363.1
11
1459
XM_017020875.1
XP_016876364.1
11
1453
XM_017020876.1
XP_016876365.1
10
2793
XM_017020877.1
XP_016876366.1
12
1628
GenBank mRNA
AF072934.1
-
9
1227
AK055086.1
-
11
1627
AK294343.1
-
10
1413
BC042196.1
-
9
1256
5198308
Back To Top
More Information
Panther Classification:
Molecular Function -
translation release factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline