Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271170.1 | NP_001258099.1 | 13 | 3916 | ||
GenBank mRNA | AY724482.1 | - | 4 | 2329 |