Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308100.1 | NP_001295029.1 | 6 | 2474 | ||
NM_001308105.1 | NP_001295034.1 | 3 | 813 | |||
NM_005460.3 | NP_005451.2 | 4 | 1029 | |||
NR_131761.1 | 4 | 954 | ||||
XM_005272138.3 | XP_005272195.1 | 4 | 952 | |||
XM_005272139.1 | XP_005272196.1 | 4 | 892 | |||
XM_006714734.2 | XP_006714797.1 | 4 | 1085 | |||
XM_011543737.2 | XP_011542039.1 | 5 | 1087 | |||
XM_011543738.2 | XP_011542040.1 | 5 | 1086 | |||
XM_011543739.1 | XP_011542041.1 | 5 | 1012 | |||
XM_011543741.2 | XP_011542043.1 | 5 | 1036 | |||
XM_011543743.2 | XP_011542045.1 | 5 | 1461 | |||
XM_011543745.1 | XP_011542047.1 | 4 | 916 | |||
XM_017010078.1 | XP_016865567.1 | 5 | 3414 | |||
XM_017010079.1 | XP_016865568.1 | 5 | 1087 | |||
XM_017010080.1 | XP_016865569.1 | 5 | 1087 | |||
XM_017010081.1 | XP_016865570.1 | 5 | 1085 | |||
XM_017010082.1 | XP_016865571.1 | 4 | 916 | |||
XM_017010084.1 | XP_016865573.1 | 5 | 1093 | |||
XM_017010086.1 | XP_016865575.1 | 5 | 1091 | |||
GenBank mRNA | AB110788.1 | - | 3 | 767 | ||
AB110789.1 | - | 3 | 767 | |||
AB110790.1 | - | 4 | 908 | |||
AF076929.1 | - | 4 | 860 | |||
AK310835.1 | - | 4 | 861 | |||
BC040552.1 | - | 6 | 2336 | 5298588 |