Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752173.3 | XP_003752221.2 | 3 | 295 | ||
XM_008773705.1 | XP_008771927.1 | 2 | 859 |
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271175.1 | NP_001258104.1 | 7 | 847 | ||
XM_006232913.2 | XP_006232975.1 | 8 | 1237 | |||
XM_006232914.2 | XP_006232976.1 | 7 | 1018 | |||
XM_006232916.2 | XP_006232978.1 | 3 | 214 | |||
XM_006232917.2 | XP_006232979.1 | 3 | 717 |