Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004297.3 | NP_004288.1 | 7 | 1438 | ||
XM_011519224.1 | XP_011517526.1 | 6 | 675 | |||
XM_017015341.1 | XP_016870830.1 | 7 | 966 | |||
GenBank mRNA | AF105201.1 | - | 7 | 1053 | ||
AF493903.1 | - | 7 | 915 | |||
AK312460.1 | - | 7 | 1438 | |||
BC027886.1 | - | 7 | 1429 | 5221889 |