Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109478.1 | NP_001102948.1 | 4, 5 | 445 | ||
GenBank mRNA | CB724692.1 | - | 3, 4 | 328 |