Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099510.1 | NP_001092980.1 | 3 | 936 |
Gene Symbol: Vom2r49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099515.1 | NP_001092985.1 | 3 | 936 |
Gene Symbol: Vom2r50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099507.1 | NP_001092977.1 | 3 | 945 |
Set Membership: |
G-protein coupled receptor |