Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007629.1 | NP_001007630.1 | 5 | 371 | ||
XM_006255480.2 | XP_006255542.1 | 4 | 335 | |||
GenBank mRNA | BC061569.1 | - | 5 | 371 | 6886638 |