Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 10, 11 | 969 | ||
XM_005270294.4 | XP_005270351.1 | 11, 12 | 1102 | |||
XM_005270295.4 | XP_005270352.1 | 10, 11 | 1056 | |||
XM_005270297.3 | XP_005270354.1 | 5, 6 | 2333 | |||
XM_005270298.4 | XP_005270355.1 | 5, 6 | 1136 | |||
XM_006718079.3 | XP_006718142.1 | 12, 13 | 1199 | |||
XM_006718080.3 | XP_006718143.1 | 6, 7 | 1230 | |||
XM_011540385.2 | XP_011538687.1 | 11, 12 | 1153 | |||
XM_011540386.2 | XP_011538688.1 | 6, 7 | 2429 | |||
XM_011540387.2 | XP_011538689.1 | 2, 3 | 4075 | |||
XM_017016969.1 | XP_016872458.1 | 11, 12 | 1120 | |||
XM_017016970.1 | XP_016872459.1 | 10, 11 | 936 | |||
GenBank mRNA | AB007878.1 | - | 8, 9 | 719 | ||
AK056469.1 | - | 9, 10 | 692 | |||
AK294454.1 | - | 5, 6 | 509 | |||
AK294736.1 | - | 5, 6 | 547 | |||
AK295314.1 | - | 5, 6 | 586 | |||
AK295660.1 | - | 5, 6 | 613 | |||
AK302015.1 | - | 8, 9 | 725 | |||
AK304828.1 | - | 5, 6 | 525 | |||
AK315983.1 | - | 5, 6 | 552 |