Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137564.1 | NP_001131036.1 | 3 | 354 | ||
GenBank mRNA | BC168768.1 | - | 3 | 354 | 6891160 | |
FQ210821.1 | - | 3 | 357 | |||
FQ211084.1 | - | 3 | 354 | |||
FQ218212.1 | - | 3 | 354 | |||
FQ218903.1 | - | 3 | 353 | |||
FQ219667.1 | - | 3 | 354 | |||
FQ221583.1 | - | 3 | 351 | |||
FQ222390.1 | - | 3 | 354 | |||
FQ222722.1 | - | 3 | 354 | |||
FQ222943.1 | - | 3 | 356 | |||
FQ228591.1 | - | 3 | 354 | |||
FQ228662.1 | - | 3 | 354 | |||
FQ229013.1 | - | 3 | 354 |
Gene Symbol: Mt1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728917.3 | XP_002728963.1 | 1 | 360 |