Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129879.2 | NP_001123351.1 | 3 | 415 | ||
XM_006253847.2 | XP_006253909.1 | 3 | 419 | |||
GenBank mRNA | BC167096.1 | - | 3 | 342 | 8362489 |