Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100366216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750725.3 | XP_003750773.1 | 6 | 1843 | ||
XM_006245340.2 | XP_006245402.1 | 6 | 3474 | |||
XM_006245341.1 | XP_006245403.1 | 6 | 1743 | |||
XM_006245342.2 | XP_006245404.1 | 7 | 2504 | |||
XM_006245343.2 | XP_006245405.1 | 5 | 2815 | |||
XM_006245344.2 | XP_006245406.1 | 5 | 2045 |