Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145838.1 | NP_001139310.1 | 4 | 654 | ||
NM_001145839.1 | NP_001139311.1 | 6 | 885 | |||
NM_014637.3 | NP_055452.3 | 6 | 885 | |||
XM_006716484.2 | XP_006716547.2 | 6 | 857 | |||
XM_011517626.1 | XP_011515928.1 | 6 | 839 | |||
XM_011517627.2 | XP_011515929.1 | 6 | 775 | |||
XM_011517628.2 | XP_011515930.1 | 5 | 992 | |||
GenBank mRNA | AK298945.1 | - | 5 | 383 | ||
AK304716.1 | - | 6 | 799 | |||
AK316202.1 | - | 5 | 425 | |||
BC036116.1 | - | 6 | 799 | 5299040 | ||
BC043498.2 | - | 6 | 784 | 6057524 | ||
BX537854.1 | - | 4 | 546 | |||
BX647507.1 | - | 6 | 804 | |||
CR456910.1 | - | 4 | 616 | |||
D13634.1 | - | 5 | 633 |