Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145839.1 | NP_001139311.1 | 3 | 341 | ||
NM_014637.3 | NP_055452.3 | 3 | 341 | |||
XM_006716484.2 | XP_006716547.2 | 3 | 313 | |||
XM_011517626.1 | XP_011515928.1 | 3 | 295 | |||
XM_011517627.2 | XP_011515929.1 | 3 | 231 | |||
GenBank mRNA | AA015794.1 | - | 3 | 273 | ||
AK298945.1 | - | 3 | 232 | |||
AK304716.1 | - | 3 | 255 | |||
AK316202.1 | - | 3 | 266 | |||
BC036116.1 | - | 3 | 255 | 5299040 | ||
BC043498.2 | - | 3 | 240 | 6057524 | ||
BX647507.1 | - | 3 | 260 | |||
CR456910.1 | - | 1 | 72 | |||
D13634.1 | - | 2 | 89 |