Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109533.1 | NP_001103003.1 | 1 | 469 | ||
GenBank mRNA | BC168175.1 | - | 1 | 430 |