Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109520.1 | NP_001102990.1 | 1 | 170 | ||
XM_006251660.2 | XP_006251722.1 | 2 | 303 |