Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127636.1 | NP_001121108.1 | 4, 5 | 448 | ||
XM_008770571.1 | XP_008768793.1 | 3, 4 | 395 | |||
GenBank mRNA | BC166528.1 | - | 4, 5 | 448 | 7189304 |