Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115037.1 | NP_001108509.1 | 3 | 662 | ||
XM_008764174.1 | XP_008762396.1 | 2 | 504 | |||
GenBank mRNA | BC158672.1 | - | 3 | 575 | 8371540 |