Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023570.3 | NP_001018864.2 | 12 | 1455 | ||
NM_001023571.3 | NP_001018865.2 | 9 | 1056 | |||
NM_001319107.1 | NP_001306036.1 | 12 | 1454 | |||
NR_134968.1 | 11 | 1344 | ||||
XM_005247911.3 | XP_005247968.1 | 12 | 1454 | |||
XM_005247912.2 | XP_005247969.1 | 11 | 1268 | |||
XM_011513335.2 | XP_011511637.1 | 10 | 1174 | |||
XM_017007537.1 | XP_016863026.1 | 11 | 1337 | |||
XM_017007538.1 | XP_016863027.1 | 8 | 1139 | |||
XM_017007539.1 | XP_016863028.1 | 9 | 1042 | |||
GenBank mRNA | AB062481.1 | - | 11 | 1338 | ||
AY714228.1 | - | 12 | 1449 | |||
AY964667.1 | - | 9 | 1006 | |||
AY964668.1 | - | 12 | 1393 | |||
BC005806.2 | - | 9 | 997 | 3834623 | ||
D25278.1 | - | 12 | 1396 |