Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_224663.7 | XP_224663.5 | 2 | 1619 |
Gene Symbol: LOC100911649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751563.3 | XP_003751611.1 | 2 | 695 | ||
XM_008771171.1 | XP_008769393.1 | 1 | 821 |
Gene Symbol: LOC685463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001063889.4 | XP_001063889.2 | 361 | |||
XM_008771169.1 | XP_008769391.1 | 487 |