Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101006.1 | NP_001094476.1 | 3 | 450 | ||
XM_006257105.2 | XP_006257167.1 | 2 | 1564 | |||
XM_006257106.2 | XP_006257168.1 | 2 | 1281 | |||
XM_006257107.2 | XP_006257169.1 | 3 | 1024 | |||
GenBank mRNA | BC081885.1 | - | 3 | 450 | 7108525 |