Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031565.2 | NP_113753.2 | 3 | 509 | ||
GenBank mRNA | EX493673.1 | - | 3 | 301 | ||
FM063764.1 | - | 3 | 509 |