Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184998.1 | NP_001171927.1 | 22 | 4523 | ||
NM_001184999.1 | NP_001171928.1 | 22 | 4514 | |||
NM_014647.3 | NP_055462.2 | 22 | 4523 | |||
XM_005255704.1 | XP_005255761.1 | 22 | 4519 | |||
XM_005255706.1 | XP_005255763.1 | 22 | 4513 | |||
XM_005255707.1 | XP_005255764.1 | 22 | 4489 | |||
XM_017023900.1 | XP_016879389.1 | 22 | 4407 | |||
XM_017023901.1 | XP_016879390.1 | 22 | 4404 | |||
XM_017023902.1 | XP_016879391.1 | 21 | 4357 | |||
XM_017023903.1 | XP_016879392.1 | 21 | 4090 | |||
XM_017023904.1 | XP_016879393.1 | 21 | 4087 | |||
XM_017023905.1 | XP_016879394.1 | 22 | 3944 | |||
XM_017023906.1 | XP_016879395.1 | 22 | 3885 | |||
GenBank mRNA | AB007890.3 | - | 20 | 3609 | ||
BC137165.1 | - | 22 | 4465 | 9021162 | ||
BC137170.1 | - | 22 | 4465 | 9021167 | ||
BC144514.1 | - | 22 | 4465 | 9053041 | ||
BC144515.1 | - | 22 | 4456 | 9053042 |