Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184998.1 | NP_001171927.1 | 21 | 4276 | ||
NM_001184999.1 | NP_001171928.1 | 21 | 4267 | |||
NM_014647.3 | NP_055462.2 | 21 | 4276 | |||
XM_005255704.1 | XP_005255761.1 | 21 | 4272 | |||
XM_005255706.1 | XP_005255763.1 | 21 | 4266 | |||
XM_005255707.1 | XP_005255764.1 | 21 | 4242 | |||
XM_017023900.1 | XP_016879389.1 | 21 | 4160 | |||
XM_017023901.1 | XP_016879390.1 | 21 | 4157 | |||
XM_017023902.1 | XP_016879391.1 | 20 | 4110 | |||
XM_017023903.1 | XP_016879392.1 | 20 | 3843 | |||
XM_017023904.1 | XP_016879393.1 | 20 | 3840 | |||
XM_017023905.1 | XP_016879394.1 | 21 | 3697 | |||
XM_017023906.1 | XP_016879395.1 | 21 | 3638 | |||
GenBank mRNA | AB007890.3 | - | 19 | 3362 | ||
BC137165.1 | - | 21 | 4218 | 9021162 | ||
BC137170.1 | - | 21 | 4218 | 9021167 | ||
BC144514.1 | - | 21 | 4218 | 9053041 | ||
BC144515.1 | - | 21 | 4209 | 9053042 |