Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001067872.5 | XP_001067872.3 | 4 | 918 | ||
XM_006248789.2 | XP_006248851.1 | 3 | 1124 | |||
XM_006248790.2 | XP_006248852.1 | 3 | 1124 | |||
XM_008769017.1 | XP_008767239.1 | 3 | 1124 |