Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053867.1 | NP_446319.1 | 3 | 215 | ||
GenBank mRNA | BC086358.1 | - | 3 | 226 | 7303675 | |
FQ210153.1 | - | 3 | 243 | |||
FQ210845.1 | - | 2 | 129 | |||
FQ217312.1 | - | 3 | 243 | |||
FQ220662.1 | - | 3 | 243 | |||
FQ220913.1 | - | 3 | 246 | |||
FQ221184.1 | - | 3 | 243 | |||
FQ221191.1 | - | 3 | 232 | |||
FQ221220.1 | - | 3 | 243 | |||
FQ221227.1 | - | 3 | 243 | |||
FQ221357.1 | - | 3 | 243 | |||
FQ221503.1 | - | 3 | 243 | |||
FQ221524.1 | - | 3 | 243 | |||
FQ221585.1 | - | 3 | 243 | |||
FQ221695.1 | - | 3 | 243 | |||
FQ221696.1 | - | 3 | 244 | |||
FQ222296.1 | - | 3 | 243 | |||
FQ222704.1 | - | 3 | 242 | |||
FQ222827.1 | - | 3 | 243 | |||
FQ223078.1 | - | 3 | 243 | |||
FQ223172.1 | - | 3 | 244 | |||
FQ223294.1 | - | 3 | 250 | |||
FQ223538.1 | - | 3 | 243 | |||
FQ223612.1 | - | 3 | 232 | |||
FQ224095.1 | - | 3 | 243 | |||
FQ224271.1 | - | 3 | 244 | |||
FQ224320.1 | - | 3 | 242 | |||
FQ224742.1 | - | 3 | 243 | |||
FQ227715.1 | - | 3 | 244 | |||
FQ227806.1 | - | 3 | 272 | |||
FQ228514.1 | - | 3 | 243 | |||
FQ228556.1 | - | 3 | 243 | |||
FQ228624.1 | - | 3 | 243 | |||
FQ228630.1 | - | 3 | 243 | |||
FQ228644.1 | - | 3 | 240 | |||
FQ229028.1 | - | 3 | 243 | |||
FQ229093.1 | - | 3 | 243 | |||
FQ229217.1 | - | 3 | 243 | |||
FQ229279.1 | - | 3 | 243 | |||
FQ229346.1 | - | 3 | 243 | |||
FQ229353.1 | - | 3 | 243 | |||
FQ230785.1 | - | 3 | 243 | |||
FQ231339.1 | - | 3 | 241 | |||
FQ231778.1 | - | 3 | 243 | |||
FQ232593.1 | - | 3 | 243 | |||
FQ232768.1 | - | 3 | 244 | |||
FQ233320.1 | - | 3 | 274 | |||
FQ233909.1 | - | 3 | 246 | |||
FQ234047.1 | - | 3 | 233 | |||
FQ234186.1 | - | 3 | 244 | |||
FQ234298.1 | - | 3 | 243 | |||
FQ234781.1 | - | 3 | 243 | |||
FQ235307.1 | - | 3 | 243 | |||
U20525.1 | - | 3 | 215 |