Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109436.2 | NP_001102906.2 | 5 | 720 | ||
GenBank mRNA | BC166995.1 | - | 5 | 720 | 9027036 |