Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304447.1 | NP_001291376.1 | 10 | 2481 | ||
NM_014653.3 | NP_055468.2 | 9 | 2650 | |||
XM_017020243.1 | XP_016875732.1 | 10 | 2935 | |||
XM_017020244.1 | XP_016875733.1 | 11 | 3096 | |||
XM_017020245.1 | XP_016875734.1 | 11 | 2593 | |||
XM_017020246.1 | XP_016875735.1 | 9 | 2324 | |||
XM_017020247.1 | XP_016875736.1 | 10 | 2467 | |||
XM_017020248.1 | XP_016875737.1 | 10 | 2939 | |||
XM_017020249.1 | XP_016875738.1 | 9 | 2796 | |||
XM_017020250.1 | XP_016875739.1 | 7 | 2350 | |||
XM_017020251.1 | XP_016875740.1 | 8 | 2161 | |||
XM_017020252.1 | XP_016875741.1 | 8 | 2639 | |||
GenBank mRNA | AB018332.2 | - | 9 | 2387 | ||
AK098069.1 | - | 1 | 182 | |||
AK293764.1 | - | 8 | 1909 | |||
BC037969.1 | - | 5 | 936 | |||
BC110330.1 | - | 9 | 2433 | 5735001 | ||
BC136687.1 | - | 8 | 2179 | 9020677 | ||
BC136688.1 | - | 8 | 2179 | 9020678 | ||
BX647283.1 | - | 10 | 2432 |