Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286184.1 | NP_001273113.1 | 2 | 754 | ||
NM_014655.3 | NP_055470.1 | 2 | 754 | |||
XM_006711657.3 | XP_006711720.1 | 3 | 1136 | |||
XM_011510180.1 | XP_011508482.1 | 4 | 1487 | |||
XM_011510181.1 | XP_011508483.1 | 3 | 1312 | |||
XM_017002904.1 | XP_016858393.1 | 4 | 1487 | |||
XM_017002905.1 | XP_016858394.1 | 3 | 1312 | |||
XM_017002906.1 | XP_016858395.1 | 3 | 1133 | |||
XM_017002907.1 | XP_016858396.1 | 2 | 602 | |||
XM_017002908.1 | XP_016858397.1 | 3 | 1312 | |||
GenBank mRNA | AB007915.3 | - | 2 | 557 | ||
AK074912.1 | - | 2 | 599 | |||
AK075002.1 | - | 2 | 574 | |||
BC008843.2 | - | 2 | 578 | 4136523 | ||
BC039854.1 | - | 2 | 557 | 6061399 |