Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077530.1 | NP_001070998.1 | 1, 2 | 183 | ||
GenBank mRNA | BC105629.1 | - | 1, 2 | 183 | 7322855 |