Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEPDC5 SIRNA Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5
Gene Aliases:
DEP.5, FFEVF
Chromosome Location:
Chr.22: 31753951 - 31907034 on Build GRCh38
UniGene ID:
Hs.435022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18632
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007188.2
NP_001007189.1
15
1249
NM_001136029.2
NP_001129501.1
14
1107
NM_001242896.1
NP_001229825.1
15
1249
NM_001242897.1
NP_001229826.1
15
1153
NM_014662.4
NP_055477.1
14
1107
NR_110988.1
15
1249
XM_005261862.1
XP_005261919.1
15
1153
XM_011530557.2
XP_011528859.1
15
1256
XM_011530558.2
XP_011528860.1
15
1256
XM_011530559.2
XP_011528861.1
15
1256
XM_011530560.2
XP_011528862.1
15
1275
XM_011530561.2
XP_011528863.1
15
1256
XM_011530562.2
XP_011528864.1
15
1256
XM_011530563.2
XP_011528865.1
15
1256
XM_011530565.2
XP_011528867.1
15
1256
XM_011530568.2
XP_011528870.1
15
1256
XM_017029109.1
XP_016884598.1
15
1153
XM_017029110.1
XP_016884599.1
15
1153
XM_017029111.1
XP_016884600.1
15
1153
XM_017029112.1
XP_016884601.1
15
1153
XM_017029113.1
XP_016884602.1
15
1256
XM_017029114.1
XP_016884603.1
15
1256
XM_017029115.1
XP_016884604.1
15
1256
GenBank mRNA
AB014545.1
-
14
1106
AJ698950.1
-
14
1102
AJ698951.1
-
14
1102
AJ704764.1
-
14
1102
AK294987.1
-
15
1125
AK300737.1
-
14
1134
BC057797.1
-
15
1189
BC136612.1
-
15
1163
9020602
BC146766.1
-
14
1106
9007209
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of TOR signaling
cellular response to amino acid starvation
intracellular signal transduction
positive regulation of GTPase activity
negative regulation of TORC1 signaling
GTPase activator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline