Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109570.1 | NP_001103040.1 | 3, 4 | 334 | ||
XM_006254884.2 | XP_006254946.1 | 3, 4 | 687 | |||
XM_006254885.2 | XP_006254947.1 | 3, 4 | 686 | |||
XM_006254886.2 | XP_006254948.1 | 3, 4 | 687 |