Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172415.1 | NP_001165886.1 | 6 | 1003 | ||
NM_004323.5 | NP_004314.5 | 6 | 1003 | |||
GenBank mRNA | AF022224.1 | - | 6 | 961 | ||
AK093238.1 | - | 3 | 1662 | |||
AK096038.1 | - | 7 | 766 | |||
AK222749.1 | - | 6 | 950 | |||
AK316577.1 | - | 6 | 929 | |||
BC001936.1 | - | 6 | 964 | 2823774 | ||
BC014774.1 | - | 6 | 813 | |||
KU177932.1 | - | 6 | 703 | |||
KU177933.1 | - | 7 | 845 | |||
U46917.1 | - | 6 | 963 | |||
Z35491.1 | - | 6 | 981 |