Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109447.1 | NP_001102917.1 | 3 | 422 | ||
XM_006251382.2 | XP_006251444.1 | 2 | 395 | |||
XM_006251383.2 | XP_006251445.1 | 2 | 2036 | |||
XM_008770365.1 | XP_008768587.1 | 1 | 117 | |||
XM_008770366.1 | XP_008768588.1 | 2 | 498 | |||
GenBank mRNA | BC087145.1 | - | 2 | 172 | 7129361 |