Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109447.1 | NP_001102917.1 | 4 | 1450 | ||
XM_006251382.2 | XP_006251444.1 | 3 | 1423 | |||
XM_006251383.2 | XP_006251445.1 | 3 | 3064 | |||
XM_008770365.1 | XP_008768587.1 | 2 | 1145 | |||
XM_008770366.1 | XP_008768588.1 | 3 | 1526 | |||
GenBank mRNA | BC087145.1 | - | 3 | 1200 | 7129361 |