Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aunip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001071529.3 | XP_001071529.1 | 2 | 391 | ||
XM_006239131.2 | XP_006239193.1 | 2 | 511 |