Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256678.1 | NP_001243607.1 | 2 | 1142 | ||
NM_001256679.1 | NP_001243608.1 | 2 | 589 | |||
NM_014672.3 | NP_055487.2 | 2 | 1142 | |||
XM_005268237.3 | XP_005268294.1 | 2 | 1041 | |||
XM_011537409.2 | XP_011535711.1 | 2 | 968 | |||
XM_011537410.2 | XP_011535712.1 | 2 | 1026 | |||
XM_017021835.1 | XP_016877324.1 | 2 | 914 | |||
XM_017021836.1 | XP_016877325.1 | 2 | 1026 | |||
XM_017021837.1 | XP_016877326.1 | 2 | 1025 | |||
GenBank mRNA | AB002389.1 | - | 2 | 892 | ||
AK301931.1 | - | 2 | 363 | |||
AK304066.1 | - | 2 | 899 | |||
BC032221.1 | - | 2 | 893 | 5206545 | ||
BC044580.1 | - | 2 | 911 | 4822711 | ||
BX161394.1 | - | 2 | 874 |