Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014674.2 | NP_055489.1 | 7 | 1393 | ||
XM_011534271.2 | XP_011532573.1 | 7 | 1587 | |||
XM_011534272.2 | XP_011532574.1 | 8 | 1386 | |||
GenBank mRNA | AK292643.1 | - | 7 | 1261 | ||
AK302065.1 | - | 7 | 862 | |||
BC019088.1 | - | 7 | 1400 | 4863779 | ||
D86967.1 | - | 7 | 1319 |