Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256584.2 | XP_006256646.1 | 4 | 3008 | ||
XM_008773024.1 | XP_008771246.1 | 3 | 2624 |