Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001068698.4 | XP_001068698.2 | 1 | 1460 |
Set Membership: |
G-protein coupled receptor |