Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012291.4 | NP_036423.4 | 31 | 6274 | ||
XM_006719705.3 | XP_006719768.1 | 31 | 6341 | |||
XM_011539024.2 | XP_011537326.1 | 31 | 6302 | |||
XM_017020253.1 | XP_016875742.1 | 31 | 6392 | |||
GenBank mRNA | AK128350.1 | - | 10 | 3733 | ||
AY455930.1 | - | 30 | 6183 | |||
D79987.1 | - | 31 | 6321 |