Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271151.1 | NP_001258080.1 | 3 | 233 | ||
GenBank mRNA | FM101799.1 | - | 2 | 105 |