Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270962.1 | NP_001257891.1 | 8 | 2017 | ||
NM_001270963.1 | NP_001257892.1 | 7 | 1960 | |||
GenBank mRNA | AB003587.1 | - | 7 | 1925 | ||
AF334791.1 | - | 7 | 1925 | |||
U75448.1 | - | 6 | 1888 |